6-Bromoimidazo[1,2-a]pyrazine-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR2598-250MG OR2598-1G
APOSOR2598-250MGEA 65.4 EUR
APOSOR2598-250MG APOSOR2598-1G
6-Bromoimidazo[1,2-a]pyrazine-2-carboxylic acid 97%
6-Bromoimidazo[1,2-a]pyrazine-2-carboxylic acid
Formel: C₇H₄BrN₃O₂
Molecular Weight: 242,0300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09414724
CAS-Nummer: 1000018-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR