STAT3 Inhibitor V, Stattic 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Stattic, Stat3 Inhibitor V, 6-Nitrobenzo[b]thiophene 1,1-dioxide, 6-​Nitro-​1,​1-​dioxide-​benzo[b]thiophene, 6-Nitro[b]thiophene-1,1-dioxide

OR26006-1G OR26006-500MG OR26006-5G
APOSOR26006-1GEA 184 EUR
APOSOR26006-1G APOSOR26006-500MG APOSOR26006-5G
STAT3 Inhibitor V, Stattic 95%
STAT3 Inhibitor V, Stattic
Formel: C₈H₅NO₄S
Molecular Weight: 211,19 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00173799
CAS-Nummer: 19983-44-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR