8-Iod-1-naphthoesäure

Lieferant: Apollo Scientific

OR26125-1G
APOSOR26125-1GEA 221 EUR
APOSOR26125-1G
8-Iod-1-naphthoesäure
8-Iod-1-naphthoesäure
Formel: C₁₁H₇IO₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 13577-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR