4-(1H-Imidazol-1-yl)phenol 99%

Lieferant: Apollo Scientific

Synonyme: 4-(1-Imidazolyl)phenol, 4-(Imidazol-1-yl)phenol

OR2614-25G OR2614-5G OR2614-1G
APOSOR2614-25GEA 130 EUR
APOSOR2614-25G APOSOR2614-5G APOSOR2614-1G
4-(1H-Imidazol-1-yl)phenol 99%
4-(1H-Imidazol-1-yl)phenol
Formel: C₉H₈N₂O
Molecular Weight: 160.18 g/mol
Schmelzpunkt: 204…207 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005281
CAS-Nummer: 10041-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR