4-Allyl-5-pyridin-2-yl-4H-1,2,4-triazole-3-thiol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Allyl-2,4-dihydro-5-(pyridin-2-yl)-3H-1,2,4-triazole-3-thione
OR26180-1G
OR26180-250MG
APOSOR26180-1GEA
149
EUR
APOSOR26180-1G
APOSOR26180-250MG
4-Allyl-5-pyridin-2-yl-4H-1,2,4-triazole-3-thiol
4-Allyl-5-pyridin-2-yl-4H-1,2,4-triazole-3-thiol
Formel:
C₁₀H₁₀N₄S Molecular Weight: 218,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03945742 CAS-Nummer: 91813-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...