2-Methoxy-1-nitronaphthalin

Lieferant: Apollo Scientific
Achtung

OR26190-1G OR26190-5G
APOSOR26190-1GEA 366 EUR
APOSOR26190-1G APOSOR26190-5G
2-Methoxy-1-nitronaphthalin
2-Methoxy-1-nitronaphthalene
Formel: C₁₁H₉NO₃
Molecular Weight: 203,2 g/mol
Schmelzpunkt: 130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00100119
CAS-Nummer: 4900-66-7
EINECS: 225-529-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR