5-Methyl-4-phenyl-1,2,3-thiadiazole 95%

Lieferant: Apollo Scientific
Achtung

OR26197-1G OR26197-250MG OR26197-5G
APOSOR26197-1GEA 65.4 EUR
APOSOR26197-1G APOSOR26197-250MG APOSOR26197-5G
5-Methyl-4-phenyl-1,2,3-thiadiazole 95%
5-Methyl-4-phenyl-1,2,3-thiadiazole
Formel: C₉H₈N₂S
Molecular Weight: 176.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00100201
CAS-Nummer: 64273-28-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR