α,α'-Dibrom-m-xylol 97+%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
m-Xyloldibromid, 1,3-Bis(brommethyl)benzol, m-Xylylendibromid
OR26233-10G
OR26233-100G
OR26233-25G
APOSOR26233-10GEA
21.9
EUR
APOSOR26233-10G
APOSOR26233-100G
APOSOR26233-25G
α,α'-Dibrom-m-xylol 97+%
α,α'-Dibrom-m-xylol
Formel:
C₈H₈Br₂ Molecular Weight: 263.96 g/mol Schmelzpunkt: 74…77 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000178 CAS-Nummer: 626-15-3 EINECS: 210-931-6 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...