2-Chloro-2,4,4-trimethylpentane

Lieferant: Apollo Scientific
Achtung

OR26275-1G OR26275-250MG OR26275-500MG
APOSOR26275-1GEA 574 EUR
APOSOR26275-1G APOSOR26275-250MG APOSOR26275-500MG
2-Chloro-2,4,4-trimethylpentane
2-Chloro-2,4,4-trimethylpentane
Formel: C₈H₁₇Cl
Lagertemperatur: Raumtemperatur
MDL: MFCD00101612
CAS-Nummer: 6111-88-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR