(1R,2S)-Fmoc-2-amino-1-cyclopentanecarboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
cis-2-Aminocyclopentanecarboxylic acid, N-FMOC protected
OR2628-1G
OR2628-5G
APOSOR2628-1GEA
171
EUR
APOSOR2628-1G
APOSOR2628-5G
(1R,2S)-Fmoc-2-amino-1-cyclopentanecarboxylic acid
(1R,2S)-Fmoc-2-amino-1-cyclopentanecarboxylic acid
Formel:
C₂₁H₂₁NO₄ Molecular Weight: 351.41 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09750531 CAS-Nummer: 352707-76-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...