2-Chlor-5-hydroxybenzoesäure 98%

Lieferant: Apollo Scientific
Achtung

OR2647-1G OR2647-5G
APOSOR2647-1GEA 102 EUR
APOSOR2647-1G APOSOR2647-5G
2-Chlor-5-hydroxybenzoesäure 98%
2-Chloro-5-hydroxybenzoic acid
Formel: C₇H₅ClO₃
Molecular Weight: 172,57 g/mol
Schmelzpunkt: 178…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04038818
CAS-Nummer: 56961-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR