2-Chlor-5-methoxybenzoesäure 99%
Lieferant: Apollo Scientific
|
Achtung
|
OR2648-1G
OR2648-5G
OR2648-25G
APOSOR2648-1GEA
21.9
EUR
APOSOR2648-1G
APOSOR2648-5G
APOSOR2648-25G
2-Chlor-5-methoxybenzoesäure 99%
2-Chlor-5-methoxybenzoesäure
Formel:
C₈H₇ClO₃ Schmelzpunkt: 170…176 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00466261 CAS-Nummer: 6280-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...