4-(4-Methoxyphenyl)-1-butanol 95%

Lieferant: Apollo Scientific

Synonyme: 4-(1-Hydroxybut-4-yl)anisole

OR2659-100G OR2659-10G OR2659-25G
APOSOR2659-100GEA 343 EUR
APOSOR2659-100G APOSOR2659-10G APOSOR2659-25G
4-(4-Methoxyphenyl)-1-butanol 95%
4-(4-Methoxyphenyl)-1-butanol
Formel: C₁₁H₁₆O₂
Molecular Weight: 180.2400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002972
CAS-Nummer: 52244-70-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR