1-Acetyl-7-amino-2,3-dihydro-(1H)-indole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Acetyl-7-aminoindoline, 1-(7-Aminoindolin-1-yl)ethanone
OR2666-1G
OR2666-5G
APOSOR2666-1GEA
422
EUR
APOSOR2666-1G
APOSOR2666-5G
1-Acetyl-7-amino-2,3-dihydro-(1H)-indole
1-Acetyl-7-amino-2,3-dihydro-(1H)-indole
Formel:
C₁₀H₁₂N₂O Molecular Weight: 176.22 g/mol Schmelzpunkt: 159…160 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00203537 CAS-Nummer: 51501-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...