1-Acetyl-6-amino-2,3-dihydro-(1H)-indole 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Acetyl-6-aminoindoline
OR2669-1G
OR2669-5G
APOSOR2669-1GEA
68.3
EUR
APOSOR2669-1G
APOSOR2669-5G
1-Acetyl-6-amino-2,3-dihydro-(1H)-indole 95+%
1-Acetyl-6-amino-2,3-dihydro-(1H)-indole
Formel:
C₁₀H₁₂N₂O Molecular Weight: 176.22 g/mol Schmelzpunkt: 182…185 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00056021 CAS-Nummer: 62368-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...