2-(4-Methyl-1,3-thiazol-5-yl)ethanol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(2-Hydroxyethyl)-4-methylthiazol, 4-Methyl-5-(2-hydroxyethyl)thiazol, 4-Methyl-5-thiazoleethanol
OR2678-100G
OR2678-25G
APOSOR2678-100GEA
26.1
EUR
APOSOR2678-100G
APOSOR2678-25G
2-(4-Methyl-1,3-thiazol-5-yl)ethanol 98%
2-(4-Methylthiazol-5-yl)ethanol
Formel:
C₆H₉NOS Molecular Weight: 143.21 g/mol Siedepunkt: 135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005339 CAS-Nummer: 137-00-8 Merck Index: 12,06208 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...