2-Methoxybenzophenon
Lieferant: Apollo Scientific
|
Achtung
|
OR26815-1G
OR26815-5G
APOSOR26815-1GEA
65.4
EUR
APOSOR26815-1G
APOSOR26815-5G
2-Methoxybenzophenon
2-Methoxybenzophenon
Formel:
C₁₄H₁₂O₂ Molecular Weight: 212,25 g/mol Siedepunkt: 149…150 °C/1mmHg Schmelzpunkt: 36…38 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017163 CAS-Nummer: 2553-04-0 EINECS: 219-857-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...