2-(4-Chlorophenoxy)-2-methylpropanoyl chloride

Lieferant: Apollo Scientific
Achtung

OR26842-1G OR26842-250MG OR26842-500MG
APOSOR26842-1GEA 0 EUR
APOSOR26842-1G APOSOR26842-250MG APOSOR26842-500MG
2-(4-Chlorophenoxy)-2-methylpropanoyl chloride
2-(4-Chlorophenoxy)-2-methylpropanoyl chloride
Formel: C₁₀H₁₀Cl₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00128244
CAS-Nummer: 5542-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR