2,3-Dihydrobenzofuran-5-acetic acid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Coumaranacetic Acid, 2,3-Dihydrobenzofuranyl-5-acetic acid

OR2687-1G OR2687-5G
APOSOR2687-1GEA 21.9 EUR
APOSOR2687-1G APOSOR2687-5G
2,3-Dihydrobenzofuran-5-acetic acid
2,3-Dihydrobenzofuran-5-acetic acid
Formel: C₁₀H₁₀O₃
Molecular Weight: 178,19 g/mol
Schmelzpunkt: 102…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01941301
CAS-Nummer: 69999-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR