2-(4-Chlorphenyl)indol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4-Chlorophenyl)-1H-indole
OR26909-1G
OR26909-5G
APOSOR26909-1GEA
34.8
EUR
APOSOR26909-1G
APOSOR26909-5G
2-(4-Chlorphenyl)indol
2-(4-Chlorophenyl)indole
Formel:
C₁₄H₁₀ClN Molecular Weight: 227,69 g/mol Schmelzpunkt: 205…207 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00047159 CAS-Nummer: 1211-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...