4-Chlor-o-phenylendiamin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chlor-1,2-phenylendiamin, 4-Chlorbenzol-1,2-diamin, 1-Chlor-3,4-diaminobenzol, 4-Chlor-1,2-diaminobenzol
OR2703-100G
OR2703-25G
OR2703-500G
APOSOR2703-100GEA
78.3
EUR
APOSOR2703-100G
APOSOR2703-25G
APOSOR2703-500G
4-Chlor-o-phenylendiamin 97%
4-Chlor-o-phenylendiamin
Formel:
C₆H₇ClN₂ Molecular Weight: 142.59 g/mol Siedepunkt: 280 Schmelzpunkt: 71…73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011691 CAS-Nummer: 95-83-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...