3-Chlor-2-thenoesäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chlorthiophen-2-carbonsäure, 3-Chlor-2-thiophencarbonsäure

OR2721-5G OR2721-1G
APOSOR2721-5GEA 31.9 EUR
APOSOR2721-5G APOSOR2721-1G
3-Chlor-2-thenoesäure 98%
3-Chlor-2-thenoesäure
Formel: C₅H₃ClO₂S
Molecular Weight: 162,6 g/mol
Schmelzpunkt: 185…190 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00043888
CAS-Nummer: 59337-89-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR