Benzamidoxim
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
N'-Hydroxybenzimidamide, Benzamide oxime
OR27223-5G
OR27223-1G
OR27223-10G
APOSOR27223-5GEA
24.8
EUR
APOSOR27223-5G
APOSOR27223-1G
APOSOR27223-10G
Benzamidoxim
Benzamidoxim
Formel:
C₇H₈N₂O Molecular Weight: 136.15 g/mol Schmelzpunkt: 60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00031485 CAS-Nummer: 613-92-3 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...