4-(Benzenesulphonyl)benzaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Phenylsulphonyl)benzaldehyde
OR2728-1G
OR2728-5G
APOSOR2728-1GEA
174
EUR
APOSOR2728-1G
APOSOR2728-5G
4-(Benzenesulphonyl)benzaldehyde
4-(Benzenesulphonyl)benzaldehyde
Formel:
C₁₃H₁₀O₃S Molecular Weight: 246,29 g/mol Schmelzpunkt: 130…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00444612 CAS-Nummer: 66-39-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...