4-Hydrazino-6-pyridin-2-yl-1,3,5-triazin-2-amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-4-hydrazino-6-pyridin-2-yl-1,3,5-triazine
OR27321-1G
OR27321-250MG
APOSOR27321-1GEA
382
EUR
APOSOR27321-1G
APOSOR27321-250MG
4-Hydrazino-6-pyridin-2-yl-1,3,5-triazin-2-amine
4-Hydrazino-6-pyridin-2-yl-1,3,5-triazin-2-amine
Formel:
C₈H₉N₇ Molecular Weight: 203,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052758 CAS-Nummer: 175204-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...