6-Methoxynicotinonitril 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Methoxypyridine-5-carbonitrile
OR2738-25G
APOSOR2738-25GEA
198
EUR
APOSOR2738-25G
6-Methoxynicotinonitril 97%
6-Methoxynicotinonitril
Formel:
C₇H₆N₂O Molecular Weight: 134,1400 g/mol Schmelzpunkt: 93…97 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084981 CAS-Nummer: 15871-85-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...