Tetrahydro-4H-thiopyran-4-on 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Tetrahydrothiopyran-4-one

OR27391-100G OR27391-25G OR27391-5G
APOSOR27391-100GEA 122 EUR
APOSOR27391-100G APOSOR27391-25G APOSOR27391-5G
Tetrahydro-4H-thiopyran-4-on 98%
Tetrahydro-4H-thiopyran-4-on
Formel: C₅H₈OS
Molecular Weight: 116,18 g/mol
Schmelzpunkt: 60…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006660
CAS-Nummer: 1072-72-6
EINECS: 214-015-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR