4-Chloro-1-methyl-1H-pyrazole-3-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR27394-1G OR27394-250MG OR27394-5G
APOSOR27394-1GEA 55.1 EUR
APOSOR27394-1G APOSOR27394-250MG APOSOR27394-5G
4-Chloro-1-methyl-1H-pyrazole-3-carbaldehyde
4-Chloro-1-methyl-1H-pyrazole-3-carbaldehyde
Formel: C₅H₅ClN₂O
Molecular Weight: 144.56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00103234
CAS-Nummer: 175204-81-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR