4-Bromo-1H-pyrazole-5-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR27395-1G
OR27395-250MG
APOSOR27395-1GEA
145
EUR
APOSOR27395-1G
APOSOR27395-250MG
4-Bromo-1H-pyrazole-5-carbaldehyde
4-Bromo-1H-pyrazole-5-carbaldehyde
Formel:
C₄H₃BrN₂O Molecular Weight: 174,99 g/mol Schmelzpunkt: 178…180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00831607 CAS-Nummer: 287917-97-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...