Methyl-6-chlornicotinat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 6-chloronicotinate
OR27398-25G
OR27398-100G
OR27398-500G
APOSOR27398-25GEA
21.9
EUR
APOSOR27398-25G
APOSOR27398-100G
APOSOR27398-500G
Methyl-6-chlornicotinat 98%
Methyl-6-chlornicotinat
Formel:
C₇H₆ClNO₂ Molecular Weight: 171,58 g/mol Schmelzpunkt: 83…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023420 CAS-Nummer: 73781-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...