5,6-Dichlornicotinamid

Lieferant: Apollo Scientific
Achtung

OR27411-1G OR27411-500MG
APOSOR27411-1GEA 261 EUR
APOSOR27411-1G APOSOR27411-500MG
5,6-Dichlornicotinamid
5,6-Dichlornicotinamid
Formel: C₆H₄Cl₂N₂O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 75291-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR