4-Amino-2,6-dichlorpyridin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Dichlor-4-pyridylamin, 2,6-Dichlorpyridin-4-amin

OR27412-25G OR27412-5G OR27412-100G
APOSOR27412-25GEA 91.5 EUR
APOSOR27412-25G APOSOR27412-5G APOSOR27412-100G
4-Amino-2,6-dichlorpyridin 98%
4-Amino-2,6-dichlorpyridin
Formel: C₅H₄Cl₂N₂
Molecular Weight: 163,01 g/mol
Schmelzpunkt: 174…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052832
CAS-Nummer: 2587-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR