4-Hydroxynicotinsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR27431-1G
OR27431-5G
OR27431-10G
APOSOR27431-1GEA
23.2
EUR
APOSOR27431-1G
APOSOR27431-5G
APOSOR27431-10G
4-Hydroxynicotinsäure 97%
4-Hydroxynicotinsäure
Formel:
C₆H₅NO₃ Schmelzpunkt: 256…259 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040286 CAS-Nummer: 609-70-1 EINECS: 210-197-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...