1,3-Dimethyluracil

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Dihydroxy-1,3-dimethylpyrimidin

OR27441-25G OR27441-5G
APOSOR27441-25GEA 24.8 EUR
APOSOR27441-25G APOSOR27441-5G
1,3-Dimethyluracil
1,3-Dimethyluracil
Formel: C₆H₈N₂O₂
Molecular Weight: 140,14 g/mol
Schmelzpunkt: 119…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038065
CAS-Nummer: 874-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR