2-Ethoxy-5-iodonicotinic acid

Lieferant: Apollo Scientific
Achtung

OR2745-1G OR2745-10G
APOSOR2745-1GEA 62.5 EUR
APOSOR2745-1G APOSOR2745-10G
2-Ethoxy-5-iodonicotinic acid
2-Ethoxy-5-iodonicotinic acid
Formel: C₈H₈INO₃
Molecular Weight: 293.06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09836165
CAS-Nummer: 335078-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR