3,5-Dichlor-4-hydroxybenzaldehyd

Lieferant: Apollo Scientific
Achtung

OR27469-1G OR27469-5G
APOSOR27469-1GEA 21.9 EUR
APOSOR27469-1G APOSOR27469-5G
3,5-Dichlor-4-hydroxybenzaldehyd
3,5-Dichloro-4-hydroxybenzaldehyde
Formel: C₇H₄Cl₂O₂
Molecular Weight: 191.01 g/mol
Schmelzpunkt: 156…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017605
CAS-Nummer: 2314-36-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR