5-Bromo-2-ethoxynicotinic acid

Lieferant: Apollo Scientific
Achtung

OR2747-1G OR2747-5G OR2747-10G
APOSOR2747-1GEA 85.7 EUR
APOSOR2747-1G APOSOR2747-5G APOSOR2747-10G
5-Bromo-2-ethoxynicotinic acid
5-Bromo-2-ethoxynicotinic acid
Formel: C₈H₈BrNO₃
Schmelzpunkt: 121…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09836166
CAS-Nummer: 393184-78-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR