Methyl 5-bromo-2-ethoxynicotinate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 5-Bromo-2-ethoxynicotinate
OR2748-1G
OR2748-5G
OR2748-250MG
APOSOR2748-1GEA
186
EUR
APOSOR2748-1G
APOSOR2748-5G
APOSOR2748-250MG
Methyl 5-bromo-2-ethoxynicotinate
Methyl-5-bromo-2-ethoxynicotinate
Formel:
C₉H₁₀BrNO₃ Molecular Weight: 260.09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09842613 CAS-Nummer: 1031927-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...