Methyl-2-(acetylamino)-5-nitrobenzoate

Lieferant: Apollo Scientific

Synonyme: Methyl 2-acetamido-5-nitrobenzoate

OR27512-1G OR27512-5G
APOSOR27512-1GEA 0 EUR
APOSOR27512-1G APOSOR27512-5G
Methyl-2-(acetylamino)-5-nitrobenzoate
Methyl-2-(acetylamino)-5-nitrobenzoate
Formel: C₁₀H₁₀N₂O₅
Molecular Weight: 238,2 g/mol
Schmelzpunkt: 172,5…177.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00157624
CAS-Nummer: 5409-45-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR