Ethyl-3-chloro-2,2-dimethylpropanoate

Lieferant: Apollo Scientific
Achtung

OR27527-1G OR27527-250MG OR27527-5G OR27527-500MG
APOSOR27527-1GEA 171 EUR
APOSOR27527-1G APOSOR27527-250MG APOSOR27527-5G APOSOR27527-500MG
Ethyl-3-chloro-2,2-dimethylpropanoate
Ethyl-3-chloro-2,2-dimethylpropanoate
Formel: C₇H₁₃ClO₂
Molecular Weight: 164,63 g/mol
Siedepunkt: 38…40 °C/0,1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00220052
CAS-Nummer: 106315-37-1
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR