2-Aminobenzolsulfonamid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Anilin-2-sulfonsäureamid, 2-Aminobenzolsulfonsäureamid

OR27532-25G OR27532-5G OR27532-1G
APOSOR27532-25GEA 105 EUR
APOSOR27532-25G APOSOR27532-5G APOSOR27532-1G
2-Aminobenzolsulfonamid 98%
2-Aminobenzolsulfonamid
Formel: C₆H₈N₂O₂S
Molecular Weight: 172,21 g/mol
Schmelzpunkt: 153…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007932
CAS-Nummer: 3306-62-5
EINECS: 221-988-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR