6-Nitro-m-anissäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Methoxy-2-nitrobenzoic acid
OR27574-100G
OR27574-25G
APOSOR27574-100GEA
46.4
EUR
APOSOR27574-100G
APOSOR27574-25G
6-Nitro-m-anissäure
6-Nitro-m-anissäure
Formel:
C₈H₇NO₅ Molecular Weight: 197,15 g/mol Schmelzpunkt: 125…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151836 CAS-Nummer: 1882-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...