6-Aminochinolin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Chinolin-6-ylamin
OR27857-1G
OR27857-5G
OR27857-10G
APOSOR27857-1GEA
21.9
EUR
APOSOR27857-1G
APOSOR27857-5G
APOSOR27857-10G
6-Aminochinolin 98%
6-Aminochinolin
Formel:
C₉H₈N₂ Molecular Weight: 144,18 g/mol Siedepunkt: 146 °C/0,3mmHg Schmelzpunkt: 115…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006803 CAS-Nummer: 580-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...