Thieno[2,3-d]pyrimidin-4(3H)-one

Lieferant: Apollo Scientific
Achtung

Synonyme: Thieno[2,3-d]pyrimidin-4(1H)-one, 3,4-Dihydro-4-oxothieno[2,3-d]pyrimidine

OR2787-1G OR2787-5G
APOSOR2787-1GEA 21.9 EUR
APOSOR2787-1G APOSOR2787-5G
Thieno[2,3-d]pyrimidin-4(3H)-one
Thieno[2,3-d]pyrimidin-4(3H)-one
Formel: C₆H₄N₂OS
Molecular Weight: 152.1738 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00706456
CAS-Nummer: 14080-50-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR