2-Brom-4'-(diethylamino)acetophenon

Lieferant: Apollo Scientific
Achtung

OR27926-1G OR27926-500MG
APOSOR27926-1GEA 0 EUR
APOSOR27926-1G APOSOR27926-500MG
2-Brom-4'-(diethylamino)acetophenon
2-Brom-4'-(diethylamino)acetophenon
Formel: C₁₂H₁₆BrNO
Schmelzpunkt: 50…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151798
CAS-Nummer: 207986-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR