2-Chlor-3-methoxypyridin

Lieferant: Apollo Scientific
Achtung

OR2800-1G OR2800-5G
APOSOR2800-1GEA 21.9 EUR
APOSOR2800-1G APOSOR2800-5G
2-Chlor-3-methoxypyridin
2-Chloro-3-methoxypyridine
Formel: C₆H₆ClNO
Molecular Weight: 143.57 g/mol
Schmelzpunkt: 46…48 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426022
CAS-Nummer: 52605-96-6
EINECS: 258-039-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR