6-Brom-2-pyridincarbonitril 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromopicolinonitrile, 6-Bromo-2-pyridinecarbonitrile

OR2804-25G OR2804-5G
APOSOR2804-25GEA 279 EUR
APOSOR2804-25G APOSOR2804-5G
6-Brom-2-pyridincarbonitril 98%
6-Brom-2-pyridincarbonitril
Formel: C₆H₃BrN₂
Molecular Weight: 183,0100 g/mol
Schmelzpunkt: 100…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00234142
CAS-Nummer: 122918-25-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR