4-Amino-2-chlorpyrimidin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chlorpyrimidin-4-ylamin
OR2806-100G
OR2806-25G
APOSOR2806-100GEA
229
EUR
APOSOR2806-100G
APOSOR2806-25G
4-Amino-2-chlorpyrimidin 98%
4-Amino-2-chloropyrimidine
Formel:
C₄H₄ClN₃ Molecular Weight: 129,55 g/mol Schmelzpunkt: 197…205 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00194051 CAS-Nummer: 7461-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...