Methyl-5-formylthiophene-3-carboxylate 98%
Lieferant: Apollo Scientific
OR2809-1G
OR2809-5G
APOSOR2809-1GEA
75.4
EUR
APOSOR2809-1G
APOSOR2809-5G
Methyl-5-formylthiophene-3-carboxylate 98%
Methyl-5-formylthiophene-3-carboxylate
Formel:
C₇H₆O₃S Molecular Weight: 170,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01859945 CAS-Nummer: 67808-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...