5-Amino-1,4-benzodioxan
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,4-Benzodioxan-5-amine, 2,3-Dihydrobenzo[b][1,4]dioxin-5-amine
OR2819-1G
OR2819-5G
APOSOR2819-1GEA
0
EUR
APOSOR2819-1G
APOSOR2819-5G
5-Amino-1,4-benzodioxan
5-Amino-1,4-benzodioxan
Formel:
C₈H₉NO₂ Molecular Weight: 151,1626 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03695459 CAS-Nummer: 16081-45-1 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...